Learn More
DuPont™ Tyvek™ IsoClean™ IC193B TS Coverall, Sterile, Double Bagged
BIOPHARMA
Product Code. 17889403
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Product Code. 17889403
Brand: DuPont™ D15560745
Only null left
Add to basket
Add to basket
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Hooded gamma-sterilized coveralls are aseptically folded and feature bound internal seams, integrated overboots with a slip-retardant sole, tunnelled elastication at wrists and ankles, elasticated waist at back, Tyvek™ covered elasticated thumb loops, and a storm flap.
  • Gamma-sterilized
  • Bound internal seams
  • Elasticated waist at back for good fit
  • Tyvek™ covered elasticated thumb loops
  • Tunneled elastication at wrists and ankles
  • Front zipper closure
  • Storm flap
  • Delivers an ideal balance of protection, durability and comfort
  • Made of high density polyethylene using a patented flash spinning process
  • Provides an inherent barrier to particles, microorganisms, and non-hazardous water-based light liquid splash
  • Breathable and low linting
  • Sterilized by gamma-irradiation
  • Folded to aid aseptic donning and packaged in an ISO class 5 cleanroom
  • Packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag
  • Packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas
  • Typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments
TRUSTED_SUSTAINABILITY
Spécification
Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Boots
Cuff Style Finger Loops
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Particle
Latex Free Yes
Model D15560
Afficher plus Afficher moins
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis