Learn More
DuPont™ Tyvek 500 Collared Laboratory Coat
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Zipper and Pockets are composed of flash spun high density polyethylene, providing an idea balance of protection, durability and comfort.
Brand: DuPont™ D13496004
54.56 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Silicon non-added
- With collar, zipper closure and 2 pockets
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
HDPE | |
Zipper | |
Chemicals | |
Stitched (internal) | |
Pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up, amongst many others |
White | |
Elastic | |
2 | |
Antistatic treatment (EN 1149-5) |