missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ Collared Laboratory Coats with Press Studs, No Pockets Product Code.: 11733123
Add to basket

DuPont™ Tyvek™ Collared Laboratory Coats with Press Studs, No Pockets

Product Code. 11733123 Shop All DuPont Products
2X-Large
Click to view available options
Size:
Large
X-Large
2X-Large

Product Code. 11733123

Brand: DuPont™ D13495953

Edge
Add to basket
Add to basket

Offers enhanced protection for body parts that are more exposed to hazardous substances. Lab coat is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.

  • Silicon non-added.
  • With 5 press stud closures.
  • Elasticated cuffs (not tunnelled).
  • Stitched internal seams.
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols.
  • Excellent barrier against fine particles and fibers (down to 1 micron in size).
  • Ultra-low-linting.
  • Antistatically treated.
  • Applications include:
    • Pharmaceutical Handling
    • Chemical Protection
    • Lead and Asbestos Abatement/Remediation
    • General Maintenance/Operations
    • Spray Painting and General Clean-up
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Resistant Lab Coat
Size 2X-Large
Gender Unisex
Material HDPE
Color White
Closure Type Press Studs
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
Style Below Hip-length
No. of Pockets None
Seam Type Stitched, Internal
Certifications/Compliance EN 1149-5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek™ Collared Laboratory Coats with Press Studs, No Pockets >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.