Learn More
DuPont™ Tyvek™ Collared Laboratory Coat with Press Studs, No Pockets
Offers enhanced protection for body parts that are more exposed to hazardous substances. Tyvek™ 500 Lab Coat with Press Studs is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
Brand: DuPont™ D13398975
226.27 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Code : NEW
Additional Details : Weight : 5.74200kg
Description
- Silicon non-added.
- With 5 press stud closures.
- Elasticated cuffs (not tunnelled).
- Stitched internal seams.
- Permeable to both air and water vapor, yet repels water-based liquids and aerosols.
- Excellent barrier against fine particles and fibers (down to 1 micron in size).
- Ultra-low-linting.
- Antistatically treated.
- Applications include:
- Pharmaceutical Handling
- Chemical Protection
- Lead and Asbestos Abatement/Remediation
- General Maintenance/Operations
- Spray Painting and General Clean-up
Specifications
Lab Coat | |
HDPE | |
Press Studs | |
Chemicals | |
None | |
EN 1149-5 |
X-Large | |
White | |
Elastic | |
Below Hip-length | |
Stitched (internal) |