Learn More
DuPont™ Tyvek 500 Trousers
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ 500 Trousers offer an excellent barrier against fine particles and fibers down to 1 micron in size.
Brand:  DuPont™ D13395741
154.98 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 11743123
  • £223.00 / Pack of 50
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Flash-spun HDPE construction
  • Elasticated waist
  • Stitched seams
  • No pockets
  • White
  • PPE Category I
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Large
Flashspun HDPE
CE Certified, EN 1149-5
Applications includes pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Chemical-resistant Trousers
White
Elastic
Internal Stitched Inseam