Learn More
DuPont™ Tyvek 500 Trousers
Product Code. 12992209 Shop All DuPont Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Each
Pack of 50
Product Code. 12992209
Brand: DuPont™ D13395741
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ 500 Trousers offer an excellent barrier against fine particles and fibers down to 1 micron in size.
  • Flash-spun HDPE construction
  • Elasticated waist
  • Stitched seams
  • No pockets
  • White
  • PPE Category I
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Product Type Chemical-resistant Trousers
Material Flashspun HDPE
Color White
Certifications/Compliance CE Certified, EN 1149-5
Waist Style Elastic
For Use With (Application) Applications includes pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Item Description Internal Stitched Inseam