Learn More
DuPont™ Tychem 2000 C Yellow Apron
113.14 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Color: Yellow
Description
- Shin length
- With neck and waist ties
- Antistatic treatment
- Applications include splash or pressurized splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | Universal |
Color | Yellow |
Finish | Polymeric barrier coating |
Certifications/Compliance | (EU) 2016/425, Category III, Type PB [3], EN 14126, EN 1149-5 |
Seam Type | Seamless |