missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem 6000 F Grey Apron
166.95 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Color: Gray
Description
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tychem™ 6000 F Aprons are made from a lightweight and durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek substrate.
- With neck and waist ties
- Antistatic treatment
- Excellent chemical permeation protection to an extensive range of chemicals
- Helps to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Type | Apron |
Size | Universal |
Material | Tychem |
Color | Gray |
Construction | Heavy-duty Tyvek substrate laminated with polymeric barrier film |
Finish | Polymeric barrier film |
Includes | Neck and waist ties |
Certifications/Compliance | (EU) 2016/425, Category III, Type PB [3], EN 14126, EN 1149-5 |
Seam Type | Seamless |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'