missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem 6000 Grey Overboot
184.62 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Color: Gray; Size: Universal
Description
- Fixation ties
- Antistatic treatment
- Sole is partially stitched
- With slip-retardant sole
- Splash-proof but not fully liquid tight
- Excellent chemical permeation protection to an extensive range of chemicals
- Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | Universal |
Material | Tychem |
Color | Gray |
Disposable | Single-use |
Sterility | Sterile |
Sole Type | Anti-slip |
Thickness | 70 μm |
Packaging Quantity | 25/Pk. |
Resistance | Fluid |
Product Type | Boot Covers |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'