missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek 500 Labo Coverall
GREENER_CHOICE

Product Code. 11743113 Shop All DuPont Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Pack of 25
This item is not returnable. View return policy

Product Code. 11743113

Brand: DuPont™ D14529842

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ CHF7 Hooded Laboratory Coveralls are robust yet lightweight with a 3-piece hood and gusset for improved fit.

  • Excellent barrier against fine particles and fibres
  • Ultra-low-linting and antistatically treated
  • Tyvek zipper and zipper flap for enhanced protection
  • Composed of flash spun high density polyethylene
  • Elasticated face, wrists and ankles as well as glued-in elastic waist
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Applications include the pharmaceutical industry, laboratories, cosmetics, optical and electronics
  • Stitched internal seams to help reduce contamination from the inside to the outside of the garment
  • Attached overshoes consisting of garment material with friction-coated sole to provide added slip resistance
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size Medium
Material Tyvek
Color White
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 5 and 6, EN 1073-2
Description Zipper flap, tunneled cuff and facial elastics
Size (Metric) Chest 92 to 100 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.