missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals U-0521, 98%

£77.10 - £254.00

Chemical Identifiers

CAS 5466-89-7
Molecular Formula C10H12O3
Synonym 3',4'-Dihydroxy-2-methylpropiophenone;
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15408719
View Documents
Thermo Scientific Chemicals
J67355.F+
20 mg
£77.10
20mg
Please to purchase this item. Need a web account? Register with us today!
15418719
View Documents
Thermo Scientific Chemicals
J67355.MC
100 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5466-89-7
C10H12O3
3',4'-Dihydroxy-2-methylpropiophenone;

Specifications

White
C10H12O3
3',4'-Dihydroxy-2-methylpropiophenone;
U-0521, 98%
VDQLKIBLTMPAHI-UHFFFAOYSA-N
1-(3,4-dihydroxyphenyl)-2-methylpropan-1-one
98%
5466-89-7
180.20
79°C to 81°C
Solid
CC(C)C(=O)C1=CC=C(O)C(O)=C1
180.20
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only