missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-U-50488 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Description
Less active enantiomer of (±)-U-50488 .
Racemate and (-)-enantiomer also available.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 405.79 |
Percent Purity | >99% |
Chemical Name or Material | (+)-U-50488 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'