missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-U-50488 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Description
Selective κ-opioid agonist. At higher concentrations blocks Na+ channels.
(-)-enantiomer and (+)-enantiomer also available.
Specifications
Specifications
Quantity | 25 mg |
Formula Weight | 405.79 |
Percent Purity | >99% |
Chemical Name or Material | (+)-U-50488 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction