Learn More
Thermo Scientific Chemicals U-73122, 95%
£38.39 - £184.00
Chemical Identifiers
CAS | 112648-68-7 |
---|---|
Molecular Formula | C29H40N2O3, C29H40N2O3 |
Synonym | 1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455189
|
Thermo Scientific Chemicals
J62898.#0 |
5 mg |
£117.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475189
|
Thermo Scientific Chemicals
J62898.MA |
10 mg |
£184.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465189
|
Thermo Scientific Chemicals
J62898.M^ |
1 mg |
£38.39
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a phospholipase C, phospholipase A2, and 5-LO inhibitor. It is also determined that in SK-N-SH neuroblastoma cells, U-73122 inhibits agonist-induced down-regulation of muscarinic receptors. In addition, it is a useful tool to investigate receptor-mediated PI turnover in signal transduction. U-73122 is a potent inhibitor of human neutrophil adhesion to biological surfaces (IC50 = 50 nM) as well as adhesion-dependent granule exocytosis and oxidative burst. U-73343 (sc-201422) is useful as a negative control for investigations of U-73122 phospholipase C antagonism and its cellular consequences.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors.Chemical Identifiers
112648-68-7 | |
C29H40N2O3, C29H40N2O3 | |
1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione |
Specifications
Off-white | |
112648-68-7 | |
464.65g/mol | |
Soluble in DMSO, ethanol, or DMF at approx. 0.5mg/ml; May require heating or overnight mixing; Insoluble in water | |
Crystalline solid | |
COC1=CC=C2[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CCC2=C1)NCCCCCCN1C(=O)C=CC1=O | |
464.65 |
Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors | |
C29H40N2O3, C29H40N2O3 | |
1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione | |
Signal Transduction Reagent | |
LUFAORPFSVMJIW-ZRJUGLEFSA-N | |
1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]amino}hexyl)-2,5-dihydro-1H-pyrrole-2,5-dione | |
≥99% |
Safety and Handling
Recommended Storage : Ambient temperatures
- U-73122
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : U-73122
RUO – Research Use Only