missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet UB-S-90

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3265700135373

 View more versions of this product

Product Code. 15907476

  • £3290.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

For indoor use only

This item is not returnable. View return policy

Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly:  drawers remain open in any position, interior of drawers completely visible – access to all containers
  • Safe: drawers fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: drawers lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Services

Five years

Specifications

Specifications

Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
600 mm
482 x 979 x 702 mm
Sliding
800 mm
UB90.080.110.060.2S
Powder-coated plain
1102 mm
Light Gray
Safety storage under bench cabinet
2 x drawer (sheet steel powder-coated), 2 x perforated insert (stainless steel)
482 mm
600 x 1102 x 800 mm
Safe and approved storage of flammable hazardous substances in working areas
702 mm
2
244.50 kg
979 mm
Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.