missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | UBAC2 |
---|---|
Concentration | 0.46 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883665
|
Proteintech
25122-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873665
|
Proteintech
25122-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
UBAC2 | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q8NBM4, Q8R1K1 | |
337867, 68889 | |
UBAC2 Fusion Protein Ag19142 | |
Primary | |
-20°C | |
UBAC2 |
0.46 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PHGDHL1, UBA domain containing 2, UBAC2 | |
UBAC2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |