missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA576144
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes ubiquitin, one of the most conserved proteins known. Ubiquitin is required for ATP-dependent, nonlysosomal intracellular protein degradation of abnormal proteins and normal proteins with a rapid turnover. Ubiquitin is covalently bound to proteins to be degraded, and presumably labels these proteins for degradation. Ubiquitin also binds to histone H2A in actively transcribed regions but does not cause histone H2A degradation, suggesting that ubiquitin is also involved in regulation of gene expression. This gene consists of three direct repeats of the ubiquitin coding sequence with no spacer sequence. Consequently, the protein is expressed as a polyubiquitin precursor with a final amino acid after the last repeat. Aberrant form of this protein has been noticed in patients with Alzheimer's and Down syndrome.Specifications
Ubiquitin C | |
Polyclonal | |
Unconjugated | |
UBC | |
2700054O04Rik; AI194771; HMG20; polyubiquitin C; polyubiquitin-C; Rps27a; TI-225; Uba52; Ubb; Ubc; Ubiquitin; ubiquitin C; Ubiquitin-related; Ubiquitin-related 1; Ubiquitin-related 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22190, 50522, 7316 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P0CG48, P0CG50, Q63429 | |
UBC | |
Synthetic peptide of human Poly-Ubiquitin. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction