missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBLCP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558859
This item is not returnable.
View return policy
Description
Immunogen sequence: SLEDVLGPPP DNDDVVNDFD IEDEVVEVEN REENLLKISR RVKEYKVEIL NPPREGKKLL VLDVDYTLFD HRSCAETG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
UBLCP1 may specifically dephosphorylate 'Ser-5' of the heptad repeats YSPTSPS in the C-terminal domain of the largest RNA polymerase II subunit.Specifications
UBLCP1 | |
Polyclonal | |
Unconjugated | |
UBLCP1 | |
4930527B16Rik; 8430435I17Rik; BC002236; CPUB1; CTD phosphatase-like with ubiquitin domain 1; CTD-like phosphatase domain-containing protein; fb33g09; MGC128319 protein; nuclear proteasome inhibitor UBLCP1; RGD1310386; ubiquitin like domain containing CTD phosphatase 1; ubiquitin-like domain containing CTD phosphatase 1; ubiquitin-like domain-containing CTD phosphatase 1; Ublcp1; wu:fb33g09; zgc:86634 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
134510, 360514, 79560 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5FWT7, Q8BGR9, Q8WVY7 | |
UBLCP1 | |
Recombinant protein corresponding to Human UBLCP1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction