missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCB 35625, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
Description
Potent chemokine CCR1 and CCR3 receptor antagonist. Inhibits MIP-1α-induced chemotaxis in CCR1 transfectants and eotaxin-induced chemotaxis in CCR3 transfectants (IC50 values are 9.57 and 93.8 nM respectively). Antagonizes CCR3-mediated entry of HIV-1 isolate 89.6 into NP-2 cells (IC50 = 57 nM).
Isomer also available.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 655.44 |
Percent Purity | >98% |
Chemical Name or Material | UCB 35625 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'