missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCHL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12384-1-AP-150UL
This item is not returnable.
View return policy
Description
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.Specifications
UCHL3 | |
Polyclonal | |
Unconjugated | |
UCHL3 | |
Ubiquitin carboxyl terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, UCHL3, UCH-L3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
498560, 50933, 7347 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15374, Q91Y78, Q9JKB1 | |
Uchl3 | |
UCHL3 Fusion Protein Ag3053 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UCHL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction