Learn More
Tocris Bioscience™ UCPH 101
Selective non-substrate EAAT1 inhibitor
Brand: Tocris Bioscience™ 3490/10
Additional Details : Weight : 0.00970kg
Description
UCPH 101 is a selective non-substrate inhibitor of EAAT1 (IC50 values are 660, >300000 and >300000 nM for EAAT1, EAAT2 and EAAT3 respectively). Also demonstrates no significant inhibition at EAAT4 or EAAT5 in a patch-clamp electrophysiology assay (at final concentration up to 10 μM).Specifications
2-Amino-5,6,7,8-tetrahydro-4-(4-methoxyphenyl)-7-(naphthalen-1-yl)-5-oxo-4H-chromene-3-carbonitrile | |
10 mg | |
C27H22N2O3 |
1118460-77-7 | |
Glutamate (EAAT) Transporter Inhibitors | |
98% |