Learn More
VELP Scientifica™ UDK 129 Kjeldahl Distillation Unit
Entry-level distillation for Kjeldahl protein determination, Non-Protein Nitrogen (NPN), volatiles and more. Simple and reliable solution with unique features.
Brand: VELP Scientifica™ F30200125
Additional Details : Weight : 32.50000kg
Beschreibung
The UDK 129 is an entry-level and reliable solution for safe distillations. The most suitable distiller for laboratories looking for a simple but efficient instrument for Kjeldahl nitrogen TKN, proteins, ammoniacal nitrogen, nitric nitrogen (Devarda), TVBN, sulfites, phenols, volatile acids, cyanides, and alcohol content.
The Kjeldahl distillation unit UDK 129 includes features that ensure efficient and reliable distillation, far beyond the expectations of an ordinary entry-level unit.
- Programmable sodium hydroxide addition
- High-precision pump ensuring constant accurate dosing of the reagent
- Exclusive titanium condenser and patented steam generator
- Intuitive interface for easy analysis settings
- Maximum safety for the operator
- Unmatched flexibility with a wide accessory range
Spezifikation
2200 W at 230 V | |
Test tubes Ø 42 x 300 mm (A00001080) Collecting flask 250 mL (10001106) Pincer for test tubes (10000247) | |
15 A | |
Corrosion-resistant technopolymer | |
CE, UKCA, Nemko, CB TEST CERTIFICATE CERTIFICAT D'ESSAI OC | |
UDK 129 Distiller without Titrator | |
50/60 Hz |
230 V | |
780 mm | |
Kjeldahl Distillation Unit | |
25 kg | |
416 x 385 x 780 mm | |
Not Autoclavable |