missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UK 14,304 tartrate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Water-soluble form of UK 14,304 (Cat. No. 0425). Full α2 adrenergic agonist. Centrally active following systemic administration in vivo.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 442.23 |
Percent Purity | >99% |
Chemical Name or Material | UK 14, 304 tartrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'