Learn More
ULBP-1 Antibody (MM0592-10K10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11986-0.025MG
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18001637
  • £153.34 / 25µg
Estimated Shipment: 01-08-2024
to see stock.
Description
Description
ULBP-1 Monoclonal specifically detects ULBP-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.
Specifications
Specifications
ULBP-1
Monoclonal
LYOPH
Flow Cytometry 1:50-1:100, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready
Q9BZM6
ULBP1
Human ULBP1
0.025 mg
Primary
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Flow Cytometry, Blocking Assay, CyTOF
MM0592-10K10
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1
Mouse
Protein G purified
RUO
80329
Human
Purified