missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-1 Antibody (MM0592-10K10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11986
Additional Details : Weight : 0.00970kg
Description
ULBP-1 Monoclonal specifically detects ULBP-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
ULBP-1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 1:50-1:100, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
Q9BZM6 | |
ULBP1 | |
Human recombinant ULBP-1 | |
0.1 mg | |
Immunology | |
80329 | |
Human | |
Purified |
Flow Cytometry, Blocking Assay, CyTOF | |
MM0592-10K10 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 |
For Research Use Only