missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ulex Europaeus (Gorse) Agglutinin I (UEA I), fluorescein (FITC)

Ulex Europaeus Agglutinin I (UEA I), fluorescein (FITC), is a bright-green UEA I-fluorophore conjugate.

Brand:  Invitrogen™ L32476

Product Code. 16648022

  • £195.00 / 2mg
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

UEA I interacts specifically with blood group H type 2 antigen, which is expressed in individuals with type O blood and to a lesser degree in individuals with type A or B blood. UEA I has been used to differentiate between angiosarcomas and epithelial tumors. However, blood group antigens are found in a variety of tissues, so better results may be obtained with the use of specific monoclonal antibodies. UEA I is also a marker for incompletely differentiated gastrin cells.

• Very bright FITC-conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: fucose, arabinose
• Inhibiting/eluting sugar: 50–100 mM L-fucose
• Blood group specificity: O (H)
• Unconjugated lectin molecular weight: 63 KDa
• Immunofluorescence working concentration: 5–20 μg/mL

UEA I-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluorescein
Gorse, furze
Approved for shipment at Room Temperature or on Wet Ice
Lectins
2 mg
When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after receipt.

Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.