Learn More
Erlab™ ULPA U16 Filter
Erlab™ ULPA U16 Filter can be use with a range of Captair Smart filtrating fume hoods and storage cabinets.
Brand: Erlab™ H01074061
911.34 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 2.00000kg
Description
- ULPA U16 filtration efficiency: 99.999995 % according to MPPS method, EN1822 standard
- For use with powders
Specifications
ULPA U16 | |
Filter | |
Powders |