missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ ULT Freezer Storage Bin

Product Code. p-7224966
Click to view available options
Dimensions (L x W x H) Exterior:
560 x 140 x 280 mm
560 x 145 x 235 mm
560 x 280 x 290 mm
610 x 180 x 220 mm
610 x 180 x 240 mm
650 x 205 x 220 mm
680 x 280 x 280 mm
685 x 280 x 280 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17295594

Brand: Wesbart™ BIN672

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Wesbart Storage Bin - A useful way of keeping loose items safe within a freezer.

  • Storage bins for ULT upright freezers
  • For storage of loose items
  • All bins are supplied with a handle or hand grip and label holder.

Specifications

Type Storage Bin
For Use With (Application) Keeping Loose Items Safe Within a Freezer
Includes Handle on One End and a Label Holder
Dimensions (L x W x H) Exterior 560 x 140 x 280 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.