missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Ultimus Bottle Top Dispenser

Combines unique functionality, user-friendliness and performance. Fisherbrand™ Ultimus Bottle Top Dispenser offers dilution, rinising, dual liquid handling and re-circulation in a single unit.

Brand:  Fisherbrand™

Additional Details : Weight : 1.33292kg

 View more versions of this product

Product Code. 15508911

  • £416.58 / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket

Description

Description

  • With dual inlet technology
  • Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
  • Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
  • Specially designed adaptor allows 360° rotation providing full flexibility
  • PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
  • Adaptors to fit most of the laboratory reagent bottles
  • Colied extension tube for refilling
  • Calibration tool provided for convenient and quick in-lab user re-calibration
 
Specifications:
  • Piston: PTFE and ETP
  • Valve housing: PFA
  • Re-Circulation valve housing: PFA
  • Valve assembly: Borosilicate glass ball and seat
  • Discharge assembly: PTFE
  • Delivery and Inlet tube: FEP
  • Compatibility: With all reagents except HF
Specifications

Specifications

0.6% (+/-0.0360)
Autoclavable at 121°C, 15psi for 10-15 min.
Borosilicate Glass
1.0 mL
Bottletop Dispenser
ISO 8655 standards
5 to 60 mL
ULT-60
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers