missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Ultimus Bottle Top Dispenser
Combines unique functionality, user-friendliness and performance. Fisherbrand™ Ultimus Bottle Top Dispenser offers dilution, rinising, dual liquid handling and re-circulation in a single unit.
£279.84 - £573.46
Specifications
Product Type | Bottletop Dispenser |
---|---|
Certifications/Compliance | ISO 8655 standards |
Material | Borosilicate Glass |
Product Code | Brand | Model | Volume (Metric) | Accuracy | Increments | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Volume (Metric) | Accuracy | Increments | Price | Quantity & Availability | ||
15558901
|
Fisherbrand™
|
ULT-10 | 1 to 10 mL | 0.6% (+/-0.060) | 0.2 mL |
£478.06
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||
15568901
|
Fisherbrand™
|
ULT-100 | 10 to 100 mL | 0.6% (+/-0.600) | 2.0 mL |
£573.46
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
||
15578901
|
Fisherbrand™
|
ULT-2.5 | 0.25 to 2.5 mL | 0.6% (+/-0.015) | 0.05 mL |
£279.84
Each |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||
15588901
|
Fisherbrand™
|
ULT-30 | 2.5 to 30 mL | 0.6% (+/-0.180) | 0.5 mL |
£416.58
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
||
15598901
|
Fisherbrand™
|
ULT-5 | 0.5 to 5 mL | 0.6% (+/-0.030) | 0.1 mL |
£313.76
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||
15508911
|
Fisherbrand™
|
ULT-60 | 5 to 60 mL | 0.6% (+/-0.0360) | 1.0 mL |
£416.58
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||
Description
- With dual inlet technology
- Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
- Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
- Specially designed adaptor allows 360° rotation providing full flexibility
- PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
- Adaptors to fit most of the laboratory reagent bottles
- Colied extension tube for refilling
- Calibration tool provided for convenient and quick in-lab user re-calibration
Specifications:
- Piston: PTFE and ETP
- Valve housing: PFA
- Re-Circulation valve housing: PFA
- Valve assembly: Borosilicate glass ball and seat
- Discharge assembly: PTFE
- Delivery and Inlet tube: FEP
- Compatibility: With all reagents except HF
Specifications
Bottletop Dispenser | |
ISO 8655 standards | |
Borosilicate Glass |