missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Ultra-Low Attachment Flasks
GREENER_CHOICE

Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.

Brand:  Corning™ 3815

 View more versions of this product

Product Code. 10710114

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 25 to 75 cm2 available growth area
  • Manufactured from optically clear virgin polystyrene
  • Ultra-Low Attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation
  • Nonpyrogenic

Order Info

Sterilized by gamma radiation; certified nonpyrogenic

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.25 oz.
HDPE
Polystyrene
Canted
6/Pk.
Ultra-low Attachment
Sterile
Vented
Orange
7.5 mL
For pipetting or scraping
24/Cs.
Rectangular
Yes
Rectangular
25 cm2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Ultra-Low Attachment Flasks > 25 cm2; Ultra-Low Attachment; Rectangular; Vent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.