missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Ultra-Low Attachment Flasks
Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Brand: Corning™ 4616
Description
- Manufactured from optically clear virgin polystyrene
- Ultra-Low Attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation
- Surface is non-cytotoxic, biologically inert, and nondegradable
- Lot numbers for ease in traceability
- 100% integrity tested
- Sterilized by gamma irradiation
- Nonpyrogenic
Specifications
0.25 oz. | |
HDPE | |
Polystyrene | |
Canted | |
6/Pk. | |
Ultra-low Attachment | |
Rectangular |
Orange | |
7.5 mL | |
For Pipetting or Scraping | |
24/Cs. | |
Rectangular | |
Sterile | |
Vented |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Ultra-Low Attachment Flasks > 25 cm2 available growth area; Rectangular Canted Neck Cell Culture Flask with Vent Cap; Ultra-Low Attachment
Spot an opportunity for improvement?Share a Content Correction