Learn More
Digi-Sense™ Ultra Low Freezer Verification Thermometer
Ultra Low Freezer Verification Thermometer - Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. Thermometers can be removed for recalibration. A magnetic attachment with pressure-sensitive backing enables the bottle to be attached to a metal wall or door.
Brand:  Digi-Sense™ 90300-69
60.25 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 15511874
  • £80.35 / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Enclosed, unbreakable plastic bottle ensures accuracy
  • Ideal for use in environmental chambers
  • Individually serial number printed on the thermometer stem
  • Non-mercury blue spirit liquid
  • NIST-traceable statement of accuracy included
  • Permanently fused markings
  • Triangular-style, anti-roll fitting
  • PFA-coated white-back glass
  • Propylene glycol filled bottle
Specifications
Specifications
Propylene Glycol
Blue Spirit
-90°C to 25°C
±1.5°C; ±2.5°C below -50°C
145 mm
PFA-coated, white-back glass ULT freezer varification thermometer
Freezers, refrigerators, incubators, ovens, environmental chambers or other environments where temperature stability is required
1°C