missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hellma Ultra-Micro Cuvette, 105.250-QS
Highly pure Suprasil™ quartz glass fluorescence cuvette with black sidewalls and a PE stopper.
Brand: Hellma 105-250-85-40
Additional Details : Weight : 0.25000kg
Description
- High-purity synthetic fused silica quartz construction
- Fluorescence cell
- UV/VIS range
- Black side walls
- PE stopper included
Specifications
100 μL | |
PE | |
PE Stopper | |
10 x 2 mm | |
45 mm |
Suprasil Quartz | |
2 mm | |
1/Pk | |
200 to 2500 nm | |
12.5 mm |