Learn More
CleanSpace™ Ultra Power System
Battery-powered, positive pressure air-purifying respirator with an IP Rating 66. Suitable for decontamination procedures and applications where there is an abundance of dust and water. Can be used for particulate and gas applications.
Brand: CleanSpace™ PAF-0070
164.43 GBP valid until 2025-01-17
Use promo code "21645" to get your promotional price.
This item is not returnable.
View return policy
Description
- IP Rating 66 – resistant to dust and liquid ingress
- Suitable for decontamination showers
- Light weight and comfortable
- AirSensit™ System for mask pressure control and breath responsive airflow
- One-button system and standby and auto-start/stop mode for quick don/doffing
- Easy to use support harness
- Altitude compensated filter blocked detection
- Long operating time: up to 8 hours and quick battery recharging: < 2 hours
- Easy and accurate flow capability check
Specifications
PAPR Assembly, No Headtop | |
PAF-0070 |
Product Content Correction
Product Title
Mask sold separately
Spot an opportunity for improvement?Share a Content Correction