missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ultra thin-bore sample tubes, 1mm ID, <3.98mm OD
For use with MKII Magnetic Susceptibility Balances
£62.50 - £296.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15431447
|
Thermo Scientific Chemicals
|
1 Each |
£62.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15476155
|
Thermo Scientific Chemicals
036152.KJ |
5 Pack | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- I.D.: 1mm
Specifications
Ultra Thin-bore Sample Tubes |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only