Learn More
Invitrogen™ UltraComp eBeads™ Plus Compensation Beads
UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry.
Brand:  Invitrogen™ 01-3333-42
249.90 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
 View more versions of this product
Product Code. 16651194
  • £357.00 / 100 tests
Explore more special offers
Description
Description
  • Binds the spherical particles for use as a single-color compensation controls.
  • Binds with mouse, rat and hamster antibodies, but also bind with rabbit and recombinant human antibodies.
  • Designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers.
  • Compensate in a manner similar to cells when used with Brilliant Violet 785/786-, Brilliant Violet 711-, Super Bright 780- and Super Bright 702-conjugated antibodies.
  • Multispecies compatibility—bind with mouse, rat, hamster, rabbit and recombinant human antibodies
  • Improved fluorophore compatibility
  • Each drop of beads contains two populations: a positive population that bind with any mouse, rat, hamster, rabbit or recombinant human antibody and a negative population that will not bind with antibody.
  • When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed.
  • This bimodal distribution can be used for single-color compensation controls to accurately setup instrumentation for multicolor flow cytometry experiments.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 x 106 beads/mL
UV-633/635 nm
100
Compensation Beads
5 μm
Mouse, rat, hamster, human, rabbit antibodies
100 tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ UltraComp eBeads™ Plus Compensation Beads > 100 Tests
RUO use only.