missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Ultrafiltration Discs, 30 kDa NMW, Biomax Polyethersulfone
30 kDa NMWL, Biomax polyethersulfone. Assemble your Amicon Stirred Cell with the membrane of your choice.
£232.00 - £510.00
Specifications
Filter Type | Ultrafiltration |
---|---|
MWCO | 30 kDa |
Filter Code | PBTK |
For Use With (Application) | Protein purification; protein concentration; protein desalting |
Housing Material | PES |
Codice del prodotto | Marca | Diameter (Metric) Filter | Filtration Area | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Diameter (Metric) Filter | Filtration Area | Prezzo | Quantità e disponibilità | ||||
10370211
|
Merck
PBTK04310 |
4.5 cm | 13.4 cm2 |
£291.00
Pack of 10 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||||
10298040
|
Merck
PBTK02510 |
2.5 cm | 4.1 cm2 |
£232.00
Pack of 10 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||||
10678451
|
Merck
PBTK06210 |
6.35 cm | 28.7 cm2 |
£455.00
Pack of 10 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||||
10054410
|
Merck
PBTK07610 |
76 mm | 41.8 cm2 |
£510.00
Pack of 10 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||||
Descrizione
30 kDa NMWL, Biomax polyethersulfone. Assemble your Amicon Stirred Cell with the membrane of your choice.
Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.
Features & Benefits:
- Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
- Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements
- Compatible with Amicon™ Stirred Cells
Applications:
Concentration, Buffer Exchange, Desalting
Specifica
Ultrafiltration | |
PBTK | |
PES |
30 kDa | |
Protein purification; protein concentration; protein desalting | |
10Pack |