missing translation for 'onlineSavingsMsg'
Learn More

Merck Ultrafiltration Discs, 300 kDa NMW, Biomax Polyethersulfone

300 kDa NMWL, Biomax polyethersulfone

Brand:  Merck PBMK04710

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10641412

  • £437.00 / Pack of 10
Estimated Shipment: 12-09-2024
to see stock.
Add to basket

Description

Description

300 kDa NMWL, Biomax polyethersulfone

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.

Features & Benefits:

  • Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
  • Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements
  • Compatible with Amicon™ Stirred Cells

Applications:
Concentration, Buffer Exchange, Desalting

Specifications

Specifications

Ultrafiltration
300000 MWCO
17.3 cm2
PES
Ultrafiltration Disc
47 mm
PES
PBMK
Protein purification; protein concentration; protein desalting
10Pack
1.85 in.
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers