missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Ultrafiltration Membrane Discs

For sample concentration in stirred cells

Brand:  Sartorius 14459--63------D

243.55 GBP valid until 2024-12-31
Use promo code "23270" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10052301

  • £284.00 / Pack of 10
Estimated Shipment: 17-09-2024
to see stock.
Add to basket

Description

Description

Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.

  • Wide range of MWCO
  • Wide range of disc diameters
  • Compatible with all stirred cells

Specifications

Specifications

Hydrosart
For sample concentration in stirred cells
180 μm
30 kDa
10/Pk.
63 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers