Learn More
Sartorius Ultrafiltration Membrane Discs
For sample concentration in stirred cells
Brand: Sartorius 14429--25------D
126.69 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.
- Wide range of MWCO
- Wide range of disc diameters
- Compatible with all stirred cells
Specifications
Hydrosart | |
For sample concentration in stirred cells | |
180 μm |
5000 | |
10/Pk. | |
25 mm |