Learn More
Sartorius Ultrafiltration Membrane Discs
For sample concentration in stirred cells
Brand:  Sartorius 14459--25------D
126.69 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10504201
  • £148.00 / Pack of 10
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.
  • Wide range of MWCO
  • Wide range of disc diameters
  • Compatible with all stirred cells
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hydrosart
For sample concentration in stirred cells
180 μm
30 kDa
10/Pk.
25 mm