missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Ultrafiltration Membrane Discs

Product Code. 10716092 Shop All Sartorius Products
Click to view available options
Diameter (Metric):
25 mm
47 mm
63 mm
76 mm
Material (Membrane):
Cellulose
PES
Polysulfone
Thickness (Metric):
120 μm
180 μm
MWCO:
1 kDa
10 kDa
100 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10716092

Brand: Sartorius 1463947D

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For sample concentration in stirred cells

Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.

  • Wide range of MWCO
  • Wide range of disc diameters
  • Compatible with all stirred cells
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Membrane Filter
Format Ultrafiltration Disc
Material PES
Material (Membrane) Polysulfone
MWCO 10 kDa
Sterility Non-sterile
For Use With (Application) For sample concentration in stirred cells
Quantity 10/Pk.
Thickness (Metric) 120 μm
Type Membrane Filter
Diameter (Metric) 47 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.