missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ DNase/RNase-Free Distilled Water
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
140.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
500 mL
10 x 500 mL
Description
UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.
Performance and Quality Testing: No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored to show compliance with current USP monograph test requirements for Water for Injection (WFI).
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Packaging | 1 Each |
Treatment(s) | Not DEPC-treated |
Quantity | 10 x 500 mL |
For Research Use Only. Not for use in diagnostic procedures.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'