Learn More
Fisherbrand™ Q125 Sonicator without Probe
Effective for standard cell disruption, DNA/RNA shearing, homogenization and many other applications. Fisherbrand™ Q125 Sonicator without Probe is a microprocessor-based, programmable ultrasonic processor.
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 4.54000kg
Description
- Programmable operation to set time for hands-free operation
- Pulse mode to prevent heat build-up in temperature-sensitive samples
- Digital amplitude/intensity control from 20-100%
- Elapsed time indicator displays duration of sonication
- Display of wattage and joules for real-time energy monitoring
- Overload protection prevents damage to circuitry if a fault occurs
- Compact design to take up less space than competitive units
- Uses lead-free components
Specifications
50/60 Hz | |
Generator, converter, power cable, converter cable, wrench set | |
125 w |
110 V | |
Q125 | |
20 kHz |