missing translation for 'onlineSavingsMsg'
Learn More

Kinematica™ Polytron™ PT2500E Desktop Homogenizer

Provides fast, reproducible preparation of most plant and animal products used for further testing and is an excellent choice for the budget-conscious laboratory which needs the versatility provided by a higher wattage motor.

Brand:  Kinematica™ 11010057

Product Code. 12799316

  • £2640.00 / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Applicable volume range from 0.05mL to 2L
  • 500W motor speed regulation from 500rpm to 30,000rpm
  • Quick coupling for dispersing aggregates
  • Easy operation with an on/off switch and speed control
  • Intelligent variable speed controls for long equipment life
  • Speed stability even with changes in viscosity
  • Easy to adjust motor height
  • Efficient sample preparation - fast dispersion saves time
  • Custom aggregates for any special application are possible
  • Interchangeable dispersing aggregates from 3mm to 25mm
  • Sleek, sound-insulated casing conceals enough power to safely and quickly handle a variety of applications
  • Ideal for general homogenization applications such as dispersion and emulsification, tissue homogenization for metabolic research and biopsy results, fast dissolution of pills, coated tablets for quality control, cell disruption, RNA/DNA isolation from tissues and much more
Specifications

Specifications

230 V
500 W
157 x 76 x 234 mm
Polytron PT-MR 2500 E
0°C to 40°C
50/60 Hz
0.05 mL to 2 L (working)
Homogenization, dispersion, emulsification, dissolution, cell disruption, RNA/DNA isolation, extraction of active substances, and much more
66 dB at 25,000 rpm, 72 dB at 30,000 rpm
1800 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kinematica™ Polytron™ PT2500E Desktop Homogenizer > Model: Polytron PT-MR 2500 E

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.