missing translation for 'onlineSavingsMsg'
Learn More
uvex Ultrasonic Safety Goggles Product Code.: 10228092
In Stock
to see stock.
Add to basket

uvex Ultrasonic Safety Goggles

Product Code. 10228092 Shop All Uvex Products
Black, Gray
Click to view available options
Frame Color:
Black, Gray
Gray, Orange

Product Code. 10228092

Brand: uvex 9302285

In Stock
to see stock.
Edge
Add to basket
Add to basket

Soft, flexible component provides complete side protection even over prescription spectacles.

  • Goggles in sporty design, also suitable as overspecs
  • Lenses can be changed
  • PC lens, clear
  • Certified to EN 166 (personal eye protection) and EN 170 (UV filters)
  • Labelling: W 166 34 B CE – 2-1.2 W 1 B KN DIN CE
  • Reliable UV 400 protection
  • High mechanical strength (B: 120 m/s)
  • Anti-fog inside, extremely scratch-resistant and chemical-resistant on the outside (uvex supravision excellence)
  • Wide panoramic field of vision
  • No pressure problems with long-term wear thanks to combination of hard and soft components
TRUSTED_SUSTAINABILITY

Specifications

Lens Type HC-AF
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Goggles
Frame Color Black, Gray
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
uvex Ultrasonic Safety Goggles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.