missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Ultrospec™ 30 Cell Density Meter

Ultrospec™ 30 Cell Density Meter for measuring density of bacteria and yeast cells in suspension.

Brand:  Biochrom 80-3008-01

Product Code. 17968480

  • £1300.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Small, portable, and easy to use, the instrument can measure growth rates of E. coli, other bacteria, and yeast
  • Create and save cell specific standard curves to read against again and again
  • Can also be used with McFarland standards
  • Intuitive touch screen for easy plug and play use
  • Battery operated and portable for use where cells are actually cultured
  • Uses LED light source in combination with an advanced fiber optics
  • Hand-held
  • Accepts 10 x 10mm cuvettes or 16 mm test tubes. 10 to 12 mm test tubes require adapter set
  • Spillages can be easily wiped from the smooth surface, and the cell compartment area can be flushed with formaldehyde or ethylene oxide to sterilize it
  • Save results onboard or export via USB flash drive
  • Compact, low-maintenance device
  • Not recommended for use with mammalian cell lines
Specifications

Specifications

CE and UKCA Marked, US and CAN Safety (ETL or equivalent)
10 x 10 mm Cuvettes, 16 mm Test Tubes
CellDensity Meter
Photodiode
170 x 210 x 50 mm
LED
Visible Spectrophotometer
600 nm
Microbiology, Bacteria and Yeast Quantification
LED, Diode Array
Absorbance 0.3 to 2.0 A
USB-C, UK, Euro, USA, China
Round
Black
USB-A, USB-C
600 nm
LCD Touchscreen
English, Chinese
40 nm
5 V
600 nm
4.3 in., 480 x 272 Pixel Resolution Backlit LCD Color Display
< ± 0.05 A, 1 A using Neutral Density Filter
± 0.02 A at 1 A om 10 mm Cuvette
100 to 240 V, 50/60 Hz, 10 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Contains internal rechargeable Li-Ion battery