missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Unassembled Convenience Kits: EPA Vials with Caps and Septa
Designed for discrete water sampling applications.
Brand: Thermo Scientific™ B7921-B
Code : 17
Additional Details : Weight : 2.32000kg
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Each carton contains shrink-wrapped vials and separately packaged caps and natural-colored PTFE/clear Silicone septa in polybags.Specifications
Amber 20mL vials, PTFE/Silicone septa and cap | |
Amber | |
20 mL | |
Discrete Water Sampling Applications |