missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC45A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15479-1-AP-20UL
This item is not returnable.
View return policy
Description
UNC45A, also named as SMAP1, acts as co-chaperone for HSP90. UNC45A prevents the stimulation of HSP90AB1 ATPase activity by AHSA1. It is a positive factor in promoting PGR function in the cell. It is necessary for proper folding of myosin and necessary for normal cell proliferation, normal myotube formation and myosin accumulation during muscle cell development. UNC45A plays a role in erythropoiesis in stroma cells in the spleen.Specifications
UNC45A | |
Polyclonal | |
Unconjugated | |
UNC45A | |
GC UNC45, GCUNC 45, GCUNC45, IRO039700, Protein unc 45 homolog A, SMAP 1, SMAP1, unc 45 homolog A (C. elegans), Unc 45A, UNC45A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101869, 308759, 55898 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q32PZ3, Q99KD5, Q9H3U1 | |
UNC45A | |
UNC45A Fusion Protein Ag7790 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |